Skip to Content
Merck
All Photos(5)

Key Documents

SAB4200515

Sigma-Aldrich

Anti-Arc antibody produced in rabbit

enhanced validation

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-Activity-regulated cytoskeleton-associated protein, Anti-Arc3.1, Anti-Arg3.1

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€469.00

€469.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€469.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€469.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~55 kDa

species reactivity

rat, mouse, human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 20 μg/mL using formalin-fixed, paraffin-embedded rat brain
western blot: 1.5-3 μg/mL using U937 cell extracts

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARC(23237)
mouse ... Arc(11838)
rat ... Arc(54323)

General description

Activity regulated cytoskeleton associated protein (Arc) is encoded by the gene mapped to human chromosome 8q24.3.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminal region of human Arc, conjugated to KLH. The corresponding sequence is identical in rat and mouse Arc.

Biochem/physiol Actions

Activity regulated cytoskeleton associated protein (Arc) plays a vital role in memory formation. It also facilitates activity-dependent plasticity of dendrites. Mutation in the gene has been associated with the development of idiopathic generalized epilepsy (IGE).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutation screening of the chromosome 8q24.3-human activity-regulated cytoskeleton-associated gene (ARC) in idiopathic generalized epilepsy.
Haug K
Molecular and Cellular Probes, 14, 255-260 (2000)
Insulin-induced expression of the activity-regulated cytoskeleton-associated gene (ARC) in human neuroblastoma cells requires p21ras, mitogen-activated protein kinase/extracellular regulated kinase and src tyrosine kinases but is protein kinase C-independent
Kremerskothen J
Neuroscience Letters, 321, 153-156 (2002)
G L Lyford et al.
Neuron, 14(2), 433-445 (1995-02-01)
Neuronal activity is an essential stimulus for induction of plasticity and normal development of the CNS. We have used differential cloning techniques to identify a novel immediate-early gene (IEG) cDNA that is rapidly induced in neurons by activity in models
Joseph Phan et al.
Scientific reports, 10(1), 13611-13611 (2020-08-14)
The two hypothalamic neuropeptides oxytocin and melanin concentrating hormone (MCH) share several physiological actions such as the control of maternal care, sexual behavior, and emotions. In this study, we uncover the role for the oxytocin-MCH signaling pathway in mood regulation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service