Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Check Cart for Availability | € 453,00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
rat, human
species reactivity (predicted by homology)
primate (based on 100% sequence homology)
technique(s)
immunohistochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FABP1(2168)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: 2.0 µg/mL of this antibody detected FABP1 in 10 µg of HepG2 cell lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| antibody form affinity isolated antibody | antibody form serum | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| species reactivity rat, human | species reactivity human, mouse | species reactivity human | species reactivity rhesus macaque, rat, mouse, human |
Still not finding the right product?
Explore all of our products under Anti-FABP1 Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



