Skip to Content
Merck
All Photos(1)

Key Documents

862231

Sigma-Aldrich

(1R,2S)-(−)-Ephedrine hydrochloride

99%

Synonym(s):

(1R,2S)-(−)-Ephedrine hydrochloride, (1R,2S)-α-[1-(Methylamino)ethyl]benzyl alcohol hydrochloride, (1R,2S)-(−)-2-Methylamino-1-phenyl-1-propanol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€120.00
100 G
€174.00

€120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€120.00
100 G
€174.00

About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · HCl
CAS Number:
Molecular Weight:
201.69
Beilstein:
3630391
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€120.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

crystals

optical activity

[α]20/D −34°, c = 4 in H2O

mp

216-220 °C (lit.)

functional group

amine
hydroxyl
phenyl

SMILES string

Cl.CN[C@@H](C)[C@H](O)c1ccccc1

InChI

1S/C10H15NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,10-12H,1-2H3;1H/t8-,10-;/m0./s1

InChI key

BALXUFOVQVENIU-GNAZCLTHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianmin Shu et al.
Journal of forensic sciences, 64(5), 1482-1485 (2019-03-06)
Ephedrine (EPH) and pseudoephedrine (PSE) were studied by micro-Raman spectroscopy and UV resonance Raman spectroscopy excited at 785 and 360 nm, respectively. Raman bands at approximately 245 and 410 cm-1 for ephedrine have apparent differences from the same bands at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service