Skip to Content
Merck
All Photos(1)

Key Documents

AB9880

Sigma-Aldrich

Anti-Huntingtin-Interacting Protein (HIP1) Antibody

Chemicon®, from rabbit

Synonym(s):

HIP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€459.00

€459.00


Estimated to ship on06 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€459.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€459.00


Estimated to ship on06 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HIP1(3092)

Specificity

Huntingtin-interacting protein 1 (HIP1). By Western blot the antibody recognizes a band at ~116 kDa on P2 microsomal fraction from embryonic rat brain.

Immunogen

Synthetic peptide

Application

Anti-Huntingtin-Interacting Protein (HIP1) Antibody detects level of Huntingtin-Interacting Protein (HIP1) & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Synapse & Synaptic Biology
Western blot: 1:500-1:1,000 on P2 microsomal fraction from embryonic rat brain.





Optimal working dilutions must be determined by end user.

Target description

116 kDa

Physical form

Format: Purified
Liquid

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service