Skip to Content
Merck
All Photos(1)

Key Documents

AB5674P

Sigma-Aldrich

Anti-Acid Sensitive Ion Channel 1 Antibody

Chemicon®, from rabbit

Synonym(s):

ASIC1, BNC1, MDEG, BNaC1a, Amiloride-sensitive Cation Channel 2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€640.00

€640.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€640.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€640.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Recognizes rat Acid Sensitive Ion Channel 1 (ASIC1). The immunogen shows no significant sequence homology with ASIC-beta or other ASICs.

Immunogen

A 20 amino acid peptide sequence near the extracellular N-terminus of rat ASIC1 (Chen et al. 1998; Waldmann et al. 1997; Garcia-Anoveros et al. 1997; Waldmann et al. 1996).

The immunogen sequence has 100% homology with human sodium channel 2 or BNaC2.

Application

Detect Acid Sensitive Ion Channel 1 using this Anti-Acid Sensitive Ion Channel 1 Antibody validated for use in ELISA & WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blot: 1-10 μg/mL using ECL.

Immunohistochemistry: not tested. It is recommended that the antibody be tried at 1-20 μg/mL on paraformaldehyde fixed tissue.

ELISA: 0.5-1 μg/mL using 1 μg/mL control peptide (Catalog number AG394) per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in phosphate buffered saline containing 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neil D Detweiler et al.
Journal of applied physiology (Bethesda, Md. : 1985), 127(2), 393-407 (2019-06-07)
Acid-sensing ion channels (ASICs) are voltage-insensitive cation channels that contribute to cellular excitability. We previously reported that ASIC1 in pulmonary artery smooth muscle cells (PASMC) contribute to pulmonary vasoreactivity and vascular remodeling during the development of chronic hypoxia (CH)-induced pulmonary
Selina M Garcia et al.
The Journal of general physiology, 155(2) (2022-12-10)
Acid-sensing ion channel 1a (ASIC1a) belongs to a novel family of proton-gated cation channels that are permeable to both Na+ and Ca2+. ASIC1a is expressed in vascular smooth muscle and endothelial cells in a variety of vascular beds, yet little
Selina M Garcia et al.
Frontiers in molecular biosciences, 9, 989809-989809 (2022-10-25)
Acid-sensing ion channel 1a (ASIC1a) is a voltage-independent, non-selective cation channel that conducts both Na+ and Ca2+. Activation of ASIC1a elicits plasma membrane depolarization and stimulates intracellular Ca2+-dependent signaling pathways in multiple cell types, including vascular smooth muscle (SM) and
Selina M Garcia et al.
PloS one, 15(7), e0236288-e0236288 (2020-07-24)
Although voltage-gated Ca2+ channels (VGCC) are a major Ca2+ entry pathway in vascular smooth muscle cells (VSMCs), several other Ca2+-influx mechanisms exist and play important roles in vasoreactivity. One of these is store-operated Ca2+ entry (SOCE), mediated by an interaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service