Skip to Content
Merck
All Photos(1)

Key Documents

SML2749

Sigma-Aldrich

iFSP1

≥98% (HPLC)

Synonym(s):

1-Amino-3-(4-methylphenyl)pyrido[1,2-a]benzimidazole-2,4-dicarbonitrile, 1-Amino-3-(p-tolyl)benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile, 1-Amino-3-p-tolyl-benzo[4,5]imidazo[1,2-a]pyridine-2,4-dicarbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€89.00
25 MG
€288.00

€89.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€89.00
25 MG
€288.00

About This Item

Empirical Formula (Hill Notation):
C20H13N5
CAS Number:
Molecular Weight:
323.35
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€89.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC1=C(C#N)C(C2=CC=C(C)C=C2)=C(C#N)C3=NC4=C(C=CC=C4)N31

InChI

1S/C20H13N5/c1-12-6-8-13(9-7-12)18-14(10-21)19(23)25-17-5-3-2-4-16(17)24-20(25)15(18)11-22/h2-9H,23H2,1H3

InChI key

FNESYDFRCQEEKA-UHFFFAOYSA-N

Biochem/physiol Actions

iFSP1 is a potent selective inhibitor of ferroptosis-suppressor-protein 1 (FSP1) that induces ferroptosis in cancer cells. iFSP1 strongly synergizes with GPX4 inhibitors to initiate ferroptosis in cancer cells.
potent selective inhibitor of ferroptosis-suppressor-protein 1 (FSP1) that induces ferroptosis in cancer cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Doll et al.
Nature, 575(7784), 693-698 (2019-10-22)
Ferroptosis is an iron-dependent form of necrotic cell death marked by oxidative damage to phospholipids1,2. To date, ferroptosis has been thought to be controlled only by the phospholipid hydroperoxide-reducing enzyme glutathione peroxidase 4 (GPX4)3,4 and radical-trapping antioxidants5,6. However, elucidation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service