Skip to Content
Merck
All Photos(1)

Key Documents

AB16301

Sigma-Aldrich

Anti-Nitric Oxide Synthase III Antibody

Chemicon®, from rabbit

Synonym(s):

NOS III, eNOS

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€327.00

€327.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1252

Request a Bulk Order

Select a Size

Change View
100 μG
€327.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€327.00


Estimated to ship on25 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1252

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOS3(4846)

Specificity

Reactive with human NOS-III (eNOS). This immunogen sequence is 100% homologous in human, bovine, rat, and mouse, but reactivity has not been tested.

Immunogen

Synthetic peptide corresponding to amino acids 596-610 of human endothelial NOS, coupled to KLH.

Application

Anti-Nitric Oxide Synthase III Antibody detects level of Nitric Oxide Synthase III & has been published & validated for use in WB & IC.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress
Western Blot: 5 μg/mL using chemiluminescent detection. Detects ~135-140kDa band in endothelial cell preparations; Higher concentrations of the antibody may be required if colorimetric detection is used. Antibody dilution will also vary with the concentration of the antigen. Immunocytochemistry: Use at 5-10 μg/mL

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25 M NaCl, pH 7.6 with 0.1% Sodium azide as a preservative.

Storage and Stability

Store at 2-8°C in undiluted aliquots for up to 6 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fahima Syeda et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 166(1), 207-214 (2013-06-04)
The ability to regulate vascular tone is an essential cardiovascular control mechanism, with nitric oxide (NO) assumed to be a ubiquitous smooth muscle relaxant. However, the literature contains reports of vasoconstrictor, vasodilator and no response to nitroergic stimulation in non-mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service