Skip to Content
Merck
All Photos(1)

Key Documents

P-164

Supelco

Phosphatidylethanol 16:0/18:1-D5 (PEth 16:0/18:1-D5) solution

100 μg/mL in methanol ((as free phosphate)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H69D5O8P · NH4
CAS Number:
Molecular Weight:
725.04
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol ((as free phosphate))

format

single component solution

storage temp.

−70°C

General description

This stable-labeled internal standard is suitable for quantitation of PEth 16:0/18:1 in LC/MS or GC/MS applications. PEth 16:0/18:1, also known as phosphatidylethanol 16:0/18:1, is a sensitive and specific biomarker for alcohol use and abuse. Since the detection window of PEth 16:0/18:1 in blood is longer than that of ethyl glucuronide (EtG) or ethyl sulfate (EtS) in urine, measurement of blood PEth 16:0/18:1 level is routinely performed in clinical toxicology and forensic testing applications such as abstinence monitoring and workplace drug testing. PEth 16:0/18:1 has also been investigated as a biomarker to confirm prenatal alcohol exposure (PAE).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service