Skip to Content
Merck
All Photos(1)

Key Documents

AB10620

Sigma-Aldrich

Anti-Cdc4 Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€433.00

€433.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€433.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€433.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FBXW7(55294)

General description

The F-box protein family is characterized by an approximately 40 amino acid motif, the F-box. F-box proteins are divided into 3 classes: FBWs containing WD-40 domains, FBLs containing leucine-rich repeats, and FBXs containing either different protein-protein interaction modules or no recognizable motifs. They constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination.
Cdc4 is one member of this family, which is also known as AGO, FBW7, FBX30 or SEL10. CDC4 binds directly to cyclin E and targets cyclin E for ubiquitin-mediated degradation. Cdc4 proteins of amino acid lengths varying from 553 aa to 707 aa in length have been reported. Cdc4 is an inhibitor of Notch signaling that targets Notch for ubiquitin-mediated degradation after a nuclear phosphorylation event.5,6 Cdc4 interacts with presenilin 1, facilitates its ubiquitination, and alters A-beta peptide production. Thus, it may be important for Alzheimer′s disease. Mutations of the CDC4 gene are detected in ovarian and breast cancer cell lines and the CDC4 gene may also be involved in the pathogenesis of human pancreatic and endometrial cancers.

Specificity

Reactivity has been confirmed with human T47D, U2-OS, MCF-7, HepG2 and HeLa cell lysates.
Recognizes CDC4.

Immunogen

Synthetic peptide derived from an internal region of the human and mouse Cdc4 (archipelago (AGO), F-box/WD-repeat protein 7, F-box and WD-40 domain protein 7, F-box protein FBW7, FBX30 and SEL-10).

Application

Anti-Cdc4 Antibody is an antibody against Cdc4 for use in WB & IP.
Research Category
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
Western Blotting: 1-3 μg/mL
Immunoprecipitation: 7 µg/reaction
Optimal working dilutions must be determined by end user.

Physical form

ImmunoAffinity Purified
This polyclonal antibody is supplied in 400 μL of phosphate buffered saline (pH 7.4) containing 0.1% sodium azide.

Storage and Stability

Store at -20°C in undiluted aliquots for 1 year from date of shipment.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Peng Wang et al.
Annals of translational medicine, 9(10), 886-886 (2021-06-25)
Colorectal cancer (CRC), one of the most common malignancies worldwide, is associated with poor survival and has a high mortality rate. Taxol is a chemotherapeutic agent that has been clinically applied as a first-line drug against diverse cancers. Yet, development
Nanae Izumi et al.
PloS one, 7(7), e41116-e41116 (2012-08-01)
Notch signaling controls fundamental aspects of angiogenic blood vessel growth including the selection of sprouting tip cells, endothelial proliferation and arterial differentiation. The E3 ubiquitin ligase Fbxw7 is part of the SCF protein complex responsible for the polyubiquitination and thereby

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service