Skip to Content
Merck
All Photos(1)

Key Documents

317505

Sigma-Aldrich

1,2-Dioctanoyl-sn-glycerol

Cell-permeable activator of protein kinase C (PKC).

Synonym(s):

1,2-Dioctanoyl-sn-glycerol, DOG, PKC Activator IV

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€80.40

€80.40


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€80.40

About This Item

Empirical Formula (Hill Notation):
C19H36O5
CAS Number:
Molecular Weight:
344.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€80.40


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

description

RTECS - RH0764500

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

solubility

chloroform: soluble
ethyl acetate: soluble

shipped in

wet ice

storage temp.

−70°C

SMILES string

O([C@H](COC(=O)CCCCCCC)CO)C(=O)CCCCCCC

InChI

1S/C19H36O5/c1-3-5-7-9-11-13-18(21)23-16-17(15-20)24-19(22)14-12-10-8-6-4-2/h17,20H,3-16H2,1-2H3/t17-/m0/s1

InChI key

ZQBULZYTDGUSSK-KRWDZBQOSA-N

General description

Cell-permeable activator of protein kinase C (PKC). Exhibits lower affinity for PKCα than for other isozymes. Shown to mimic the effects of tumor-promoting phorbol diesters on mitogenesis and epidermal growth factor binding and action in intact cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Protein kinase C
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Analysis Note

Single spot by TLC

Other Notes

Ferro, T.J., et al. 1993. Am. J. Physiol. 264, L7.
MacEwan, D.J., et al. 1993. Eur. J. Pharmacol. 246, 9.
Davis, R.J. 1985. J. Biol. Chem.260, 1562.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service