Skip to Content
Merck
All Photos(1)

Key Documents

AB1060

Sigma-Aldrich

Anti-Rubella Antibody

Chemicon®, from goat

Synonym(s):

German Measles

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€585.00

€585.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€585.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€585.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

goat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
inhibition assay: suitable (hemagglutination)
western blot: suitable

shipped in

dry ice

Specificity

Rubella virus, purified virons. No cross-reactivity with Vero cells.

Immunogen

Strain HPV77.

Application

Detect Rubella using this Anti-Rubella Antibody validated for use in ELISA, IF, WB, HI.
Hemagglutination inhibition: >1:512

Indirect immunofluorescence: >1:2,560

ELISA

Immunoblotting; unboiled samples in 1mM DTT and 2% SDS only.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
IgG fraction (95% pure). In PBS (0.01 M, pH 7.2) with 0.1% sodium azide.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service