Skip to Content
Merck
All Photos(1)

Key Documents

8.57010

Sigma-Aldrich

Cleavage Reagent for DNA Synthesis

20% DEA in MeCN, Novabiochem®

Synonym(s):

20% DEA in MeCN (v/v), Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
€135.00

€135.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
500 ML
€135.00

About This Item

UNSPSC Code:
41116006
NACRES:
NA.22

€135.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

reaction suitability

reaction type: Aminolysis
reagent type: Lewis base

General description

Pre-mixed ready-to-use Oxidizer Reagent
  • 857010.0500: Compatible with Cytiva ÄKTA DNA Synthesisers. Stand alone from Instrument (GL-45 Neck Finish)

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

36.0 °F

Flash Point(C)

2.2 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service