Skip to Content
Merck
All Photos(1)

Key Documents

8.56087

Sigma-Aldrich

Cysteamine 4-methoxytrityl resin

Novabiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€174.00
5 G
€696.00
25 G
€2,790.00

€174.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€174.00
5 G
€696.00
25 G
€2,790.00

About This Item

UNSPSC Code:
12352116
NACRES:
NA.22

€174.00


Estimated to ship on24 April 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

Resin for the synthesis of peptidyl-aminoethylthiols by Fmoc SPPS

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Linkage

Replaces: 01-64-0086

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.20 - 1.30 mmol/g
Swelling Volume (in DMF): lot specific result
Teh polymer matrix is copoly (styrene-1% DVB), 200 - 400 mesh.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service