Skip to Content
Merck

Skip To

8.55006

MBHA resin HL (100-200 mesh) . HCl

Novabiochem®

Synonym(s):

4-Methylbenzhydrylamine resin HL (100-200 mesh) hydrochloride salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.22
UNSPSC Code:
13111023
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

High load resin for the synthesis of peptide amides by Boc SPPS [1,2]. Carboxylic acids or amino acids can be attached using standard methods of amide bond formation. Cleavage of the carboxamides from this resin can be effected by treatment with HF or TFMSA.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.80 - 1.40 mmol/g
Swelling Volume (in CH₂Cl₂): lot specific result
The polyer matrix is copoly (styrene-1 % DVB), 100 - 200 mesh

Other Notes

Replaces: 01-64-0042

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
855000855011855020
description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

description

Novabiochem®

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

functional group

amine

functional group

amine

functional group

-

functional group

amine

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters