Skip to Content
Merck
All Photos(2)

Key Documents

8.55000

Sigma-Aldrich

MBHA resin LL (100-200 mesh). HCl

Novabiochem®

Synonym(s):

4-Methylbenzhydrylamine resin LL (100-200 mesh) hydrochloride salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€81.90
25 G
€308.00
100 G
€726.00

€81.90


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€81.90
25 G
€308.00
100 G
€726.00

About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

€81.90


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Boc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-30°C

General description

Standard resin for the synthesis of peptide amides by Boc SPPS [1,2]. Carboxylic acids or amino acids can be attached using standard methods of amide bond formation. Cleavage of the carboxamides from this resin can be affected by treatment with HF or TFMSA.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins
Cleavage and Deprotection Protocols for Boc SPPS
Base Resins for SPPS

Linkage

Replaces: 01-64-0006

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.60 - 0.90 mmol/g
Swelling Volume (in DMF): lot specific result

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Anhydrous HF is the preferred reagent for peptide cleavage from Boc-based resins, versatile and effective for various peptide synthesis.

Questions

  1. I just wanted to know the porosity of the particle

    1 answer
    1. The particle porosity of this product is not determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service