Skip to Content
Merck
All Photos(1)

Documents

8.52373

Sigma-Aldrich

Fmoc-Cys(STmp)-OH

for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Cys(STmp)-OH, N-α-Fmoc-S-2,4,6-trimethoxyphenylthio-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27NO7S2
CAS Number:
Molecular Weight:
541.64
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Cys(STmp)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

thiol

storage temp.

15-25°C

General description

Fmoc-Cys(STmp)-OH is a novel new tool for the regioselective synthesis of multiple disulfide bridged peptides by Fmoc SPPS. The STmp group is stable to piperidine but is extremely easy to remove by mild thiolysis. Albericio has reported removing four STmp groups on the solid phase with only three 5 minute treatments of 0.1 M N-methylmorpholine (NMM) in DMF containing 5% DTT.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Cysteine-Containing Peptides

Literature references:
[1] T. M. Postma, et al. (2012) Org. Lett., 14, 5468.
[2] T. M. Postma & F. Albericio (2013) Org. Lett., 15, 616.

Application

Applications of Fmoc-Cys(STmp)-OH:
  • Synthesis of insulin analogs by regiospecific disulfide bond formation.
  • A review on step-wise introduction of disulfide bonds.
  • Synthesis of human insulin-like peptide 6.

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Assay (HPLC, area%): ≥ 94.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.05 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Trimethoxyphenylthio as a highly labile replacement for tert-butylthio cysteine protection in Fmoc solid phase synthesis
T. M. Postma, et al.,
Organic Letters, 14, 5468-5468 (2012)
Synthesis of Four-Disulfide Insulin Analogs via Sequential Disulfide Bond Formation
Fangzhou Wu, et al.
The Journal of Organic Chemistry, 82, 3506-3506 (2017)
N-Chlorosuccinimide, an efficient reagent for on-resin disulfide formation in solid-phase peptide synthesis
T. M. Postma & F. Albericio
Organic Letters, 15, 616-616 (2013)
Chemical Synthesis of Human Insulin-Like Peptide-6
Chemistry?A European Journal , 22, 9777-9777 (2016)
Stepwise Construction of Disulfides in Peptides
H Rongjun, et al.,
Chembiochem, 21, 1101-1101 (2020)

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Protocols

Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service