Skip to Content
Merck
All Photos(1)

Key Documents

8.41027

Sigma-Aldrich

2-Aminoheptane

for synthesis

Synonym(s):

2-Aminoheptane, 2-Heptylamine, 1-Methylhexylamine

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€35.70
25 ML
€45.40
100 ML
€177.00

€35.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 ML
€35.70
25 ML
€45.40
100 ML
€177.00

About This Item

Empirical Formula (Hill Notation):
C7H17N
CAS Number:
Molecular Weight:
115.22
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-655-5
NACRES:
NA.22

€35.70


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

pH

11.45 (20 °C, 10 g/L in H2O)

bp

142-144 °C/1013 hPa

transition temp

flash point 54 °C

density

0.768 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NC(CCCCC)C

InChI

1S/C7H17N/c1-3-4-5-6-7(2)8/h7H,3-6,8H2,1-2H3

InChI key

VSRBKQFNFZQRBM-UHFFFAOYSA-N

Application

  • Concepts for New Rapid Simple HPLC Method for Quantification of Fosfomycin Trometamol: This study utilized 2-aminoheptane in HPLC methods to quantify fosfomycin trometamol, showcasing its utility in pharmaceutical analysis (M Piponski et al., 2022).
  • Multi-step biocatalytic strategies for chiral amino alcohol synthesis: Discusses the synthesis of various chiral amines including 2-aminoheptane, highlighting its role in creating complex organic molecules (MF Villegas-Torres et al., 2015).
  • Determination of the Dissociation Constants (pKa) of Eight Amines: This research focuses on the pKa values of amines including 2-aminoheptane, which is crucial for understanding its chemical behavior in various environments (VSPV Alluri et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service