Skip to Content
Merck
All Photos(1)

Key Documents

8.21249

Sigma-Aldrich

9-Methylanthracene

for synthesis

Synonym(s):

9-Methylanthracene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€40.70
5 G
€131.00

€40.70


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€40.70
5 G
€131.00

About This Item

Empirical Formula (Hill Notation):
C15H12
CAS Number:
Molecular Weight:
192.26
MDL number:
UNSPSC Code:
12352005
EC Index Number:
212-299-7
NACRES:
NA.22

€40.70


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥97.0% (GC)

form

crystals

bp

196-197 °C/16 hPa

mp

76-79 °C

solubility

0.0003 mg/L

density

1.06 g/cm3

storage temp.

2-30°C

SMILES string

Cc1c2c(cc3c1cccc3)cccc2

InChI

1S/C15H12/c1-11-14-8-4-2-6-12(14)10-13-7-3-5-9-15(11)13/h2-10H,1H3

InChI key

CPGPAVAKSZHMBP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 76 °C
Melting range (upper value): ≤ 79 °C
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service