Skip to Content
Merck
All Photos(1)

Key Documents

8.20990

Sigma-Aldrich

1-Phenyldodecane

for synthesis

Synonym(s):

1-Phenyldodecane, Dodecyl benzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€158.00
100 ML
€458.00

€158.00


Estimated to ship on29 May 2025



Select a Size

Change View
25 ML
€158.00
100 ML
€458.00

About This Item

Linear Formula:
C6H5(CH2)11CH3
CAS Number:
Molecular Weight:
246.43
MDL number:
UNSPSC Code:
12352005
EC Index Number:
204-591-8
NACRES:
NA.22

€158.00


Estimated to ship on29 May 2025


vapor pressure

0.1 hPa ( 50 °C)

Quality Level

Assay

≥99.0% (GC)

bp

275-300 °C/1013 hPa

mp

3 °C

transition temp

flash point 124 °C

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(CCCCCC)CCCCCc1ccccc1

InChI

1S/C18H30/c1-2-3-4-5-6-7-8-9-10-12-15-18-16-13-11-14-17-18/h11,13-14,16-17H,2-10,12,15H2,1H3

InChI key

KWKXNDCHNDYVRT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.854 - 0.856
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service