Skip to Content
Merck
All Photos(1)

Key Documents

8.20934

Sigma-Aldrich

Triethyl orthoacetate

for synthesis

Synonym(s):

Triethyl orthoacetate, Orthoacetic acid triethyl ester, 1,1,1-Triethoxyethane

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
€26.10
100 ML
€44.60
500 ML
€99.20

€26.10


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
50 ML
€26.10
100 ML
€44.60
500 ML
€99.20

About This Item

Linear Formula:
CH3C(OC2H5)3
CAS Number:
Molecular Weight:
162.23
MDL number:
UNSPSC Code:
12352112
EC Index Number:
201-112-4
NACRES:
NA.22

€26.10


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

3.4 hPa ( 20 °C)

Quality Level

Assay

≥96.0% (GC)

form

liquid

autoignition temp.

455 °C

expl. lim.

0.9-7.0 % (v/v)

bp

142 °C/1013 hPa

transition temp

flash point 39 °C

density

0.8888 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OCC)(OCC)C)CC

InChI

1S/C8H18O3/c1-5-9-8(4,10-6-2)11-7-3/h5-7H2,1-4H3

InChI key

NDQXKKFRNOPRDW-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 96.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.888 - 0.892
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - Non-equilibrium method

Flash Point(C)

39 °C - Non-equilibrium method


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service