Skip to Content
Merck
All Photos(2)

Key Documents

8.14689

Sigma-Aldrich

Platinum(II) acetylacetonate

50% Pt, catalyst for synthesis

Synonym(s):

Platinum(II) acetylacetonate (50% Pt), Platinum bis(acetylacetonate), Bis(acetylacetonato) platinum

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€250.00

€250.00


Estimated to ship on26 May 2025



Select a Size

Change View
1 G
€250.00

About This Item

Empirical Formula (Hill Notation):
C10H14O4Pt
CAS Number:
Molecular Weight:
393.29
MDL number:
UNSPSC Code:
12352103
EC Index Number:
239-223-5
NACRES:
NA.22

€250.00


Estimated to ship on26 May 2025


Product Name

Platinum(II) acetylacetonate (50% Pt), catalyst for synthesis

Quality Level

potency

>500 mg/kg LD50, oral (Rat)

mp

250-252 °C

storage temp.

2-30°C

SMILES string

[Pt+2].O=C([C-H]C(=O)C)C.O=C([C-H]C(=O)C)C

InChI

1S/2C5H7O2.Pt/c2*1-4(6)3-5(2)7;/h2*3H,1-2H3;/q2*-1;+2

InChI key

TUOYDQBYTHSWNN-UHFFFAOYSA-N

Analysis Note

Assay (of Pt): 47.0 - 53.0 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service