Skip to Content
Merck
All Photos(1)

Key Documents

8.20613

Sigma-Aldrich

Guanidinium thiocyanate

for synthesis

Synonym(s):

Guanidinium thiocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€115.00
1 KG
€309.00

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
250 G
€115.00
1 KG
€309.00

About This Item

Linear Formula:
H2NC(NH)NH2 · HSCN
CAS Number:
Molecular Weight:
118.16
MDL number:
UNSPSC Code:
12352107
EC Index Number:
209-812-1
NACRES:
NA.22

€115.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

form

solid

pH

4.8-6.0 (20 °C, 1420 g/L in H2O, saturated solution)

mp

118 °C

solubility

1420 g/L

density

1.29 g/cm3 at 20 °C

bulk density

630 kg/m3

storage temp.

2-30°C

SMILES string

[S-]C#N.[N+H3]C(=N)N

InChI

1S/CH5N3.CHNS/c2-1(3)4;2-1-3/h(H5,2,3,4);3H

InChI key

ZJYYHGLJYGJLLN-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 115 °C
Melting range (upper value): ≤ 120 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service