Skip to Content
Merck
All Photos(2)

Key Documents

8.20530

Sigma-Aldrich

N,N′-Diphenylbenzidine

for synthesis

Synonym(s):

N,N′-Diphenylbenzidine, N,N′-Diphenyl-4,4′-diaminobiphenyl, N,N′-Diphenyl-[1,1′-biphenyl]-4,4′-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€54.80
10 G
€106.00

€54.80


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€54.80
10 G
€106.00

About This Item

Linear Formula:
4-(C6H5NH)C6H4C6H4-4-(NHC6H5)
CAS Number:
Molecular Weight:
336.43
MDL number:
UNSPSC Code:
12352116
EC Index Number:
208-521-7
NACRES:
NA.22

€54.80


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥96.0% (HPLC)

form

powder

mp

245-250 °C

storage temp.

2-30°C

SMILES string

N(c2ccc(cc2)c3ccc(cc3)Nc4ccccc4)c1ccccc1

InChI

1S/C24H20N2/c1-3-7-21(8-4-1)25-23-15-11-19(12-16-23)20-13-17-24(18-14-20)26-22-9-5-2-6-10-22/h1-18,25-26H

InChI key

FDRNXKXKFNHNCA-UHFFFAOYSA-N

Application


  • Direct electrochemical dimerization of N, N′-diphenylbenzidine: The research focuses on the electrochemical process to produce dimerized forms of N,N′-diphenylbenzidine, which are important in developing new materials for electronic applications (Shahparast et al., 2019).

  • Enhancement of structure and optical dispersion properties of N,N′-Bis(3-methylphenyl)-N,N′-diphenylbenzidine thin films: This paper explores the impact of UV irradiation on the optical and structural properties of thin films made from N,N′-diphenylbenzidine derivatives, important for optical applications (Abdel-Khalek et al., 2021).

Analysis Note

Assay (HPLC, area%): ≥ 96.0 % (a/a)
Melting range (lower value): ≥ 245 °C
Melting range (upper value): ≤ 251 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service