Skip to Content
Merck
All Photos(1)

Key Documents

8.14889

Sigma-Aldrich

4-Chlorophenyl-2-hydroxyethyl sulfide

for synthesis

Synonym(s):

4-Chlorophenyl-2-hydroxyethyl sulfide

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€169.00

€169.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€169.00

About This Item

CAS Number:
Molecular Weight:
188.68
MDL number:
UNSPSC Code:
12352104
EC Index Number:
236-657-7
NACRES:
NA.22

€169.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

form

liquid

bp

140 °C/1013 hPa

density

1.278 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

S(CCO)c1ccc(cc1)Cl

InChI

1S/C8H9ClOS/c9-7-1-3-8(4-2-7)11-6-5-10/h1-4,10H,5-6H2

InChI key

HHIWVLZQIJUIPJ-UHFFFAOYSA-N

General description

for synthesis

Application

4-Chlorophenyl-2-hydroxyethyl sulfide for synthesis. CAS 13457-98-2, molar mass 188.68 g/mol.

Analysis Note

Assay (GC, area%)≥ 98.0 % (a/a)Identity (IR)passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service