Skip to Content
Merck
All Photos(1)

Key Documents

8.20409

Sigma-Aldrich

Dibenzothiophene

for synthesis

Synonym(s):

Dibenzothiophene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€66.10

€66.10


Estimated to ship on15 April 2025



Select a Size

Change View
25 G
€66.10

About This Item

Empirical Formula (Hill Notation):
C12H8S
CAS Number:
Molecular Weight:
184.26
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-072-9
NACRES:
NA.22

€66.10


Estimated to ship on15 April 2025


Quality Level

Assay

≥98.0% (GC)

autoignition temp.

>450 °C

pH

7 (20 °C, 50 g/L in H2O, slurry)

bp

331-333 °C/1013 hPa

mp

95-98 °C

transition temp

flash point 170 °C

solubility

0.0015 g/L (Lit.)

storage temp.

2-30°C

SMILES string

[s]1c2c(c3c1cccc3)cccc2

InChI

1S/C12H8S/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1-8H

InChI key

IYYZUPMFVPLQIF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 96 °C
Melting range (upper value): ≤ 99 °C
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service