Skip to Content
Merck
All Photos(1)

Key Documents

8.20351

Sigma-Aldrich

Crotonoyl chloride

(mixture of cis- and trans isomers) for synthesis

Synonym(s):

Crotonoyl chloride, Crotonic acid chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€98.40

€98.40


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€98.40

About This Item

Linear Formula:
CH3CH=CHCOCl
CAS Number:
Molecular Weight:
104.53
MDL number:
UNSPSC Code:
12352106
EC Index Number:
234-010-3
NACRES:
NA.22

€98.40


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥95.0% (GC)

form

liquid

transition temp

flash point 40.5 °C

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(=O)\C=C\C

InChI

1S/C4H5ClO/c1-2-3-4(5)6/h2-3H,1H3/b3-2+

InChI key

RJUIDDKTATZJFE-NSCUHMNNSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.080 - 1.090
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F

Flash Point(C)

40.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service