Skip to Content
Merck
All Photos(1)

Key Documents

8.18650

Sigma-Aldrich

Adipic acid

for synthesis

Synonym(s):

Adipic acid, Hexanedioic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€25.70
1 KG
€38.00

€25.70


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€25.70
1 KG
€38.00

About This Item

Empirical Formula (Hill Notation):
C6H10O4
CAS Number:
Molecular Weight:
146.14
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-673-3
NACRES:
NA.22

€25.70


Estimated to ship on17 May 2025


Request a Bulk Order

vapor pressure

0.097 hPa ( 18.5 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

crystals

autoignition temp.

405 °C

potency

5700 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

pH

2.7 (25 °C, 23 g/L in H2O)

mp

150.9 °C

transition temp

flash point 196 °C

density

1.36 g/cm3 at 25 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCC(=O)[O-].[H+].[H+]

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Biodepolymerization of Polyamide Fibers Using Yarrowia lipolytica as Whole-Cell Biocatalyst: This study explores the synthesis of Polyamide-6,6 by polycondensation of two initial monomeric units adipic acid and hexamethylenediamine (FS Buarque et al., 2024).
  • Interfacial Hydrogen Bond Effect Between CeO2 and g-C3N4 Boosts Conversion to Adipic Acid from Aerobic Oxidation of Cyclohexane: This research highlights how the interfacial hydrogen bond effect between catalysts can improve the aerobic oxidation of cyclohexane to adipic acid, a significant step towards sustainable adipic acid production (K Li et al., 2024).

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 150 °C
Melting range (upper value): ≤ 154 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service