Adipic acid
Synonym(s):
Hexanedioic acid, Adipic acid
Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
Products
Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing