Skip to Content
Merck
All Photos(1)

Key Documents

8.18320

Sigma-Aldrich

Tetra-tert-butyl orthotitanate

for synthesis

Synonym(s):

Tetra-tert-butyl orthotitanate, tert-Butyl titanate(IV), Orthotitanic acid tetra-tert-butyl ester, Titanium tetra-tert-butylate, Titanic acid tetra-tert-butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€110.00

€110.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€110.00

About This Item

Linear Formula:
Ti(OC(CH3)3)4
CAS Number:
Molecular Weight:
340.32
MDL number:
UNSPSC Code:
12352103
EC Index Number:
221-412-9
NACRES:
NA.22

€110.00


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

43 hPa ( 136 °C)

Quality Level

form

liquid

bp

160-165 °C/20 hPa

transition temp

flash point 47.2 °C

density

0.88 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Ti](OC(C)(C)C)(OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

InChI

1S/4C4H9O.Ti/c4*1-4(2,3)5;/h4*1-3H3;/q4*-1;+4

InChI key

GRWPYGBKJYICOO-UHFFFAOYSA-N

Analysis Note

Assay (ex Ti): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 0.888 - 0.894
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

117.0 °F - closed cup

Flash Point(C)

47.2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service