Skip to Content
Merck
All Photos(2)

Key Documents

217190

Sigma-Aldrich

Gadolinium(III) nitrate hexahydrate

crystals and lumps, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€200.00

€200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€200.00

About This Item

Linear Formula:
Gd(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
451.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystals and lumps

€200.00


Check Cart for Availability

Request a Bulk Order

Assay

99.999% trace metals basis

form

crystals and lumps

reaction suitability

reagent type: catalyst
core: gadolinium

impurities

≤20.0 ppm Trace Rare Earth Analysis

mp

91 °C (lit.)

density

2.406 g/mL at 25 °C

SMILES string

[Gd+3].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Gd.3NO3.6H2O/c;3*2-1(3)4;;;;;;/h;;;;6*1H2/q+3;3*-1;;;;;;

InChI key

XWFVFZQEDMDSET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service