Skip to Content
Merck
All Photos(2)

Key Documents

8.18191

Sigma-Aldrich

Bis(2-ethylhexyl) amine

for synthesis

Synonym(s):

Bis(2-ethylhexyl) amine, Diisooctylamine, Di-(2-ethylhexyl)-amin

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€50.10

€50.10


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€50.10

About This Item

Linear Formula:
[CH3(CH2)3CH(C2H5)CH2]2NH
CAS Number:
Molecular Weight:
241.46
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-372-4
NACRES:
NA.22

€50.10


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

0.0023 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

230 °C

potency

1008 mg/kg LD50, oral (Rat)
956 mg/kg LD50, skin (Rabbit)

expl. lim.

0.6-3.7 % (v/v)

pH

>7 (20 °C in H2O)

bp

157-159 °C/27 hPa

mp

<-60 °C

transition temp

flash point 130 °C

solubility

0.014 g/L

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CC(CCCC)CC)CC(CCCC)CC

InChI

1S/C16H35N/c1-5-9-11-15(7-3)13-17-14-16(8-4)12-10-6-2/h15-17H,5-14H2,1-4H3

InChI key

SAIKULLUBZKPDA-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.803 - 0.805
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

266.0 °F - Pensky-Martens closed cup

Flash Point(C)

130 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service