Skip to Content
Merck
All Photos(1)

Key Documents

E29508

Sigma-Aldrich

2-Ethyl-1-hexylamine

98%

Synonym(s):

β-Ethylhexylamine, 1-Amino-2-ethylhexane, 2-Ethyl-1-aminohexane, 2-Ethyl-1-hexanamine, 2-Ethylhexanamine, 2-Ethylhexylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€27.80
1 L
€101.00

€27.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€27.80
1 L
€101.00

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CH2NH2
CAS Number:
Molecular Weight:
129.24
Beilstein:
1209249
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€27.80


Check Cart for Availability

Request a Bulk Order

vapor pressure

1.2 mmHg ( 20 °C)

Quality Level

Assay

98%

form

liquid

autoignition temp.

563 °F

refractive index

n20/D 1.431 (lit.)

bp

169 °C (lit.)

mp

−76 °C (lit.)

density

0.789 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)CN

InChI

1S/C8H19N/c1-3-5-6-8(4-2)7-9/h8H,3-7,9H2,1-2H3

InChI key

LTHNHFOGQMKPOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Ethyl-1-hexylamine can be used as a reactant to synthesize:
  • Carbonyl compounds through oxidative deamination by using N-tert-butylphenylsulfinimidoyl chloride.[1]
  • Nitriles via Ru-catalyzed oxidation in the presence of molecular oxygen.[2]
  • Alkyl isothiocyanates by reacting with carbon disulfide in the presence of a base.[3]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the water content present in 2-Ethyl-1-hexylamine.

    1 answer
    1. Unfortunately, the water content of this product has not been determined.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service