Skip to Content
Merck
All Photos(1)

Key Documents

8.14724

Sigma-Aldrich

Isoamyl butyrate

for synthesis

Synonym(s):

Isoamyl butyrate, Butyric acid (3-methylbutyl ester)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€82.90

€82.90


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€82.90

About This Item

Linear Formula:
CH3CH2CH2COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
158.24
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-380-8
NACRES:
NA.22

€82.90


Estimated to ship on24 May 2025


Request a Bulk Order

vapor pressure

1.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>5000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

mp

-73 °C

transition temp

flash point 59 °C

solubility

0.5 g/L

density

0.88 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCC(C)C)C(=O)CCC

InChI

1S/C9H18O2/c1-4-5-9(10)11-7-6-8(2)3/h8H,4-7H2,1-3H3

InChI key

PQLMXFQTAMDXIZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.860 - 0.864
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service