Skip to Content
Merck
All Photos(1)

Key Documents

05549

Supelco

trans-2,cis-6-Nonadienal

analytical standard

Synonym(s):

Violet leaf aldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€65.40

€65.40


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 MG
€65.40

About This Item

Linear Formula:
C2H5CH=CHCH2CH2CH=CHCHO
CAS Number:
Molecular Weight:
138.21
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

€65.40


Estimated to ship on29 May 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥94.5% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.472-1.480
n20/D 1.474 (lit.)

bp

94-95 °C/18 mmHg (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)C(\[H])=C(/[H])CC\C([H])=C(\[H])CC

InChI

1S/C9H14O/c1-2-3-4-5-6-7-8-9-10/h3-4,7-9H,2,5-6H2,1H3/b4-3-,8-7+

InChI key

HZYHMHHBBBSGHB-ODYTWBPASA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

181.4 °F

Flash Point(C)

83 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Böttcher et al.
Food chemistry, 169, 492-498 (2014-09-23)
The aim of the present study was to evaluate the off-flavour masking potential of pea dextrin (PD) in emulsions rich in ω-3 and ω-6-fatty acids in comparison with maltodextrin (MD) and 2-hydroxypropyl-β-cyclodextrin (HPBCD). After optimisation of the homogenisation procedure, stable
Laure Saint-Lary et al.
Chemistry & biodiversity, 11(6), 843-860 (2014-06-18)
Absolutes isolated from Viola odorata leaves, valuable materials for the flavor and fragrance industry, were studied. Violets are mainly cultivated in France and Egypt and extracted locally. The absolutes of the two origins showed different olfactory profiles both in top

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service