810967
MOD–U–CYTE VIII Media and Antisera Diluent
19-21 g/dL protein (Biuret), Cholesterol 400-500mg/dL, USA origin, Vesicular Stomatitis Virus: None detected, Bluetongue: None detected, Blood banking: suitable, media additive: suitable, Serology: suitable
About This Item
Recommended Products
Product Name
MOD–U–CYTE VIII Media and Antisera Diluent, pack of 50 mL
description
0.1% sodium azide as preservtive
Contains sodium azide less than 0.1 g/dL
Quality Level
Assay
400-500 mg/dL cholesterol basis
form
solution
shelf life
12 mo.
purified by
heat shock fractionation
packaging
pack of 50 mL
origin
USA origin
concentration
19-21 g/dL protein (Biuret)
technique(s)
blood typing: suitable
dilution
(Immunohematology)
(Serology)
(media additive)
impurities
Tested for IgG
pH
7.2-7.4
foreign activity
Bluetongue, none detected
Microbial Content tested by USP
Vesicular Stomatitis Virus, none detected
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Probumin® BSA is a commonly used bovine serum albumin for buffers, non-specific blocking, protein stabilization and other diagnostic applications. Given the diverse uses of BSA, here are some frequently asked questions and answers about the molecule, formats, methods, and applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service