Skip to Content
Merck
All Photos(1)

Key Documents

8.08735

Sigma-Aldrich

Tripropyltin chloride

for synthesis

Synonym(s):

Tripropyltin chloride, Chlorotripropyltin, Tripropylchlorotin, Chlorotripropylstannane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2)3SnCl
CAS Number:
Molecular Weight:
283.43
MDL number:
UNSPSC Code:
12352103
EC Index Number:
218-910-3
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

bp

123 °C/17.3 hPa

mp

-23 °C

transition temp

flash point 95 °C

density

1.294-1.300 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/3C3H7.Cl.Sn/c3*1-3-2;;/h3*1,3H2,2H3;;

InChI key

ANLXQSDUWZBRRH-UHFFFAOYSA-N

Analysis Note

Assay (ex Sn): ≥ 98.0 %
Density (d 20 °C/ 4 °C): 1.294 - 1.300
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

203.0 °F

Flash Point(C)

95 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service