Skip to Content
Merck
All Photos(1)

Key Documents

615633

Sigma-Aldrich

Tributyltin chloride-d27

98 atom % D, 96% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€510.00

€510.00


Check Cart for Availability

Request more information

Select a Size

Change View
100 MG
€510.00

About This Item

Linear Formula:
[CD3(CD2)3]3SnCl
CAS Number:
Molecular Weight:
352.67
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€510.00


Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

96% (CP)

bp

171-173 °C/25 mmHg (lit.)

mass shift

M+27

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[Sn](Cl)(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H]

InChI

1S/3C4H9.ClH.Sn/c3*1-3-4-2;;/h3*1,3-4H2,2H3;1H;/q;;;;+1/p-1/i3*1D2,2D3,3D2,4D2;;

InChI key

GCTFWCDSFPMHHS-FMYBXEBSSA-M

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1B - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawid Kucharski et al.
Molecules (Basel, Switzerland), 25(3) (2020-02-06)
A Quick, Easy, Cheap, Effective, Rugged, and Safe (QuEChERS) extraction method combined with Liquid Chromatography-Tandem Mass Spectrometry (LC-MS/MS) for determination of organotin compounds (OTC) has been newly developed. The novel analytical method was validated and the quality of the results
Manuel Gadogbe et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 54(9), 884-890 (2019-05-07)
Exposure to tin in the general US population is near ubiquitous, as determined using urinary tin levels measured by inductively coupled plasma mass spectrometry (ICP-MS). Urinary tin levels are associated with chronic health outcomes, such as diabetes; however, it is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service