Skip to Content
Merck
All Photos(2)

Documents

8.08249

Sigma-Aldrich

Triethylene glycol dimethyl ether

(stabilised) for synthesis

Synonym(s):

1,2-Bis(2-methoxyethoxy)ethane, Dimethyltriglycol, NSC 66400, Triglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)3CH3
CAS Number:
Molecular Weight:
178.23
Beilstein:
1700630
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-977-3
NACRES:
NA.22

vapor density

>4.7 (vs air)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

375 °F

potency

5390 mg/kg LD50, oral (Rat)

expl. lim.

0.7 % (v/v)

refractive index

n20/D 1.423 (lit.)

pH

7

bp

216 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 106 °C

solubility

soluble >1000 g/L

density

0.986 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

COCCOCCOCCOC

InChI

1S/C8H18O4/c1-9-3-5-11-7-8-12-6-4-10-2/h3-8H2,1-2H3

InChI key

YFNKIDBQEZZDLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.985 - 0.989
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service