Skip to Content
Merck
All Photos(1)

Key Documents

8.06845

Sigma-Aldrich

Nonylamine

for synthesis

Synonym(s):

Nonylamine, 1-Aminononane

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€134.00

€134.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 ML
€134.00

About This Item

Empirical Formula (Hill Notation):
C9H21N
CAS Number:
Molecular Weight:
143.27
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-945-9
NACRES:
NA.22

€134.00


Estimated to ship on25 May 2025


Request a Bulk Order

vapor pressure

0.37 hPa ( 25 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

385 °C

pH

>7 (20 °C in H2O, undiluted)

bp

202 °C/1013 hPa

mp

-1 °C

transition temp

flash point 63 °C

density

0.79 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Cl.NCCCCCCCCC

InChI

1S/C9H21N.ClH/c1-2-3-4-5-6-7-8-9-10;/h2-10H2,1H3;1H

InChI key

IMGHCWMUWVHYCO-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.787 - 0.792
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service