Skip to Content
Merck
All Photos(1)

Key Documents

8.03527

Sigma-Aldrich

Dodecylamine

for synthesis

Synonym(s):

Dodecylamine, Laurylamine, 1-Aminododecan

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€40.40
500 G
€74.60

€40.40


Estimated to ship on30 March 2025


Request a Bulk Order

Select a Size

Change View
100 G
€40.40
500 G
€74.60

About This Item

Linear Formula:
CH3(CH2)11NH2
CAS Number:
Molecular Weight:
185.35
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-690-6
NACRES:
NA.22
form:
solid
Assay:
≥97.0% (GC)

€40.40


Estimated to ship on30 March 2025


Request a Bulk Order

vapor pressure

<2 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

solid

potency

1020 mg/kg LD50, oral (Rat)

bp

247-249 °C/1013 hPa

mp

25-28 °C

transition temp

flash point >110 °C

density

0.80 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCCCCCCCCCCC

InChI

1S/C12H27N/c1-2-3-4-5-6-7-8-9-10-11-12-13/h2-13H2,1H3

InChI key

JRBPAEWTRLWTQC-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range: ≥ 25 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service