Skip to Content
Merck
All Photos(1)

Key Documents

8.06169

Sigma-Aldrich

N-Methylformanilide

for synthesis

Synonym(s):

N-Methylformanilide, Formic acid N-methylanilide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€73.50

€73.50


Estimated to ship on15 May 2025


Request a Bulk Order

Select a Size

Change View
250 ML
€73.50

About This Item

Linear Formula:
HCON(CH3)C6H5
CAS Number:
Molecular Weight:
135.16
MDL number:
UNSPSC Code:
12352111
EC Index Number:
202-262-3
NACRES:
NA.22

€73.50


Estimated to ship on15 May 2025


Request a Bulk Order

vapor pressure

2.66 Pa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

pH

3.5-5.5 (20 °C in H2O, saturated aqueous solution)

bp

243-244 °C/1013 hPa

mp

8-13 °C

transition temp

flash point 126 °C

density

1.095 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(C)(c1ccccc1)C=O

InChI

1S/C8H9NO/c1-9(7-10)8-5-3-2-4-6-8/h2-7H,1H3

InChI key

JIKUXBYRTXDNIY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.094 - 1.096
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service