Skip to Content
Merck
All Photos(1)

Key Documents

8.00974

Sigma-Aldrich

Allyl alcohol

for synthesis

Synonym(s):

Allyl alcohol, 2-Propene-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€55.90
1 L
€163.00

€55.90


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€55.90
1 L
€163.00

About This Item

CAS Number:
Molecular Weight:
58.07
MDL number:
UNSPSC Code:
12352203
EC Index Number:
203-470-7
NACRES:
NA.22

€55.90


Estimated to ship on28 May 2025


Request a Bulk Order

vapor pressure

24.3 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

375 °C

potency

64 mg/kg LD50, oral (Rat)

expl. lim.

2.5-18 % (v/v)

bp

97 °C/1013 hPa

mp

-129 °C

transition temp

flash point 22 °C

density

0.85 g/cm3 at 20 °C

SMILES string

OCC=C

InChI

1S/C3H6O/c1-2-3-4/h2,4H,1,3H2

InChI key

XXROGKLTLUQVRX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for synthesis

Application

Allyl alcohol for synthesis. CAS No. 107-18-6, EC Number 203-470-7.

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)0.851 - 0.854Water (K. F.)≤ 0.20 %Identity (IR)passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service