Skip to Content
Merck
All Photos(1)

Key Documents

8.04771

Sigma-Aldrich

Iodine monochloride

for synthesis

Synonym(s):

Iodine monochloride, Iodine chloride, Chlorine iodide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€82.20
250 G
€159.00

€82.20


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€82.20
250 G
€159.00

About This Item

Empirical Formula (Hill Notation):
ClI
CAS Number:
Molecular Weight:
162.36
MDL number:
UNSPSC Code:
12352307
EC Index Number:
232-236-7
NACRES:
NA.22

€82.20


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

liquid

bp

97 °C/1013 hPa (decomposition)

mp

14-27 °C

density

3.1 g/cm3 at 29 °C

storage temp.

2-30°C

SMILES string

ICl

InChI

1S/ClI/c1-2

InChI key

QZRGKCOWNLSUDK-UHFFFAOYSA-N

Application

  • Structural organization in the trimethylamine iodine monochloride complex: This research highlights the use of iodine monochloride as a reactant in the organic synthesis of trimethylamine iodine monochloride complex. (Marshall et al., 2017).
  • Ultrasonic-assisted synthesis of flavones: A study exploring the synthesis of flavones using iodine monochloride under ultrasonic irradiation, providing an improved method for chemical synthesis. (Lahyani and Trabelsi, 2016).
  • Reactivity and applications in synthetic approaches: This review discusses the reactivity of iodine monochloride and its applications in synthetic chemistry, highlighting its versatility. (Day et al., 2021).

Analysis Note

Assay (iodometric): ≥ 97.0 %
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service