Skip to Content
Merck
All Photos(1)

Key Documents

359122

Sigma-Aldrich

Iodine trichloride

97%

Synonym(s):

Trichloroiodine

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€112.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€112.00

About This Item

Linear Formula:
ICl3
CAS Number:
Molecular Weight:
233.26
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

€112.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

storage temp.

−20°C

SMILES string

Cl[I](Cl)Cl

InChI

1S/Cl3I/c1-4(2)3

InChI key

PAWIVBWALDNUJP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iodine trichloride (ICl3) is an inorganic compound that is commonly used as an oxidizing agent in organic synthesis. It is also used in the halogenation of organic compounds, and oxidative cleavage of carbon-carbon bonds.

Application

Iodine trichloride can be used:     
  • In the halogenation of aromatic compounds to prepare corresponding chlorinated derivatives.      
  • To oxidize primary and secondary alcohols to aldehydes and ketones.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service