Skip to Content
Merck
All Photos(1)

Key Documents

8.03544

Sigma-Aldrich

Dioctyltin oxide

for synthesis

Synonym(s):

Dioctyltin oxide, Dioctyloxostannane

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€51.20
100 G
€141.00

€51.20


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
25 G
€51.20
100 G
€141.00

About This Item

Empirical Formula (Hill Notation):
C16H34OSn
CAS Number:
Molecular Weight:
361.15
MDL number:
UNSPSC Code:
12352103
EC Index Number:
212-791-1
NACRES:
NA.22

€51.20


Estimated to ship on26 May 2025


Request a Bulk Order

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

form

powder

autoignition temp.

>100 °C

potency

2500 mg/kg LD50, oral (Rat)

bp

230 °C/1013 hPa

transition temp

flash point 70 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[Sn](=O)(CCCCCCCC)CCCCCCCC

InChI

1S/2C8H17.O.Sn/c2*1-3-5-7-8-6-4-2;;/h2*1,3-8H2,2H3;;

InChI key

LQRUPWUPINJLMU-UHFFFAOYSA-N

Analysis Note

Assay (ex Sn): ≥ 97.0 %
Identity (IR): passes test

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service