Skip to Content
Merck
All Photos(1)

Key Documents

8.22203

Sigma-Aldrich

Hexadecylamine

for synthesis

Synonym(s):

Hexadecylamine, 1-Aminohexadecan, Cetylamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€35.50
500 G
€98.40

€35.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€35.50
500 G
€98.40

About This Item

Linear Formula:
CH3(CH2)15NH2
CAS Number:
Molecular Weight:
241.46
MDL number:
UNSPSC Code:
12352116
EC Index Number:
205-596-8
NACRES:
NA.22

€35.50


Estimated to ship on30 May 2025


Request a Bulk Order

vapor pressure

1.3 hPa ( 20 °C)

Quality Level

Assay

≥94.0% (GC)

form

lumps

bp

330 °C/1013 hPa

mp

43-46 °C

transition temp

flash point 140 °C

density

0.94 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NCCCCCCCCCCCCCCCC

InChI

1S/C16H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-17H2,1H3

InChI key

FJLUATLTXUNBOT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 94.0 % (a/a)
Melting range (lower value): ≥ 44 °C
Melting range (upper value): ≤ 48 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

285.8 °F - closed cup

Flash Point(C)

141 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service