Skip to Content
Merck
All Photos(2)

Key Documents

8.02506

Sigma-Aldrich

(+)-Cinchonine

for resolution of racemates for synthesis

Synonym(s):

(+)-Cinchonine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€76.00
100 G
€204.00

€76.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€76.00
100 G
€204.00

About This Item

Empirical Formula (Hill Notation):
C19H22N2O
CAS Number:
Molecular Weight:
294.39
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-234-6
NACRES:
NA.22

€76.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

pH

9.0 (20 °C, 0.25 g/L in H2O, as aqueous solution)

mp

258-264 °C

solubility

0.25 g/L

bulk density

130 kg/m3

storage temp.

2-30°C

SMILES string

N21[C@H](C[C@@H]([C@H](C2)C=C)CC1)[C@@H](O)c3c4c(ncc3)cccc4

InChI

1S/C19H22N2O/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17/h2-7,9,13-14,18-19,22H,1,8,10-12H2/t13-,14-,18+,19-/m0/s1

InChI key

KMPWYEUPVWOPIM-QAMTZSDWSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Spec. rotation [α²0/D (c=0.5 in methanol): +218.0 - +222.0 °
Identity (IR): passes test
Contains about 15 % Dihydrocinchonine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service