Skip to Content
Merck
All Photos(1)

Key Documents

298352

Sigma-Aldrich

(1S)-(−)-2,10-Camphorsultam

98%

Synonym(s):

(−)-10,2-Camphorsultam, (−)-exo-10,2-Bornanesultam, (1S,5R)-10,10-Dimethyl-3-thia-4-azatricyclo[5.2.1.01,5]decane 3,3-dioxide, [3aS-(3aα,6α,7aβ)]-hexahydro-8,8-dimethyl-3H-3a,6-methano-2,1-benzisothiazole-2,2-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€120.00

€120.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€120.00

About This Item

Empirical Formula (Hill Notation):
C10H17NO2S
CAS Number:
Molecular Weight:
215.31
Beilstein:
83811
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€120.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

optical activity

[α]19/D −32°, c = 5 in chloroform

mp

181-183 °C (lit.)

SMILES string

[H][C@@]12CC[C@]3(CS(=O)(=O)N[C@@H]3C1)C2(C)C

InChI

1S/C10H17NO2S/c1-9(2)7-3-4-10(9)6-14(12,13)11-8(10)5-7/h7-8,11H,3-6H2,1-2H3/t7-,8-,10-/m1/s1

InChI key

DPJYJNYYDJOJNO-NQMVMOMDSA-N

General description

(1S,2R,4R)-(−)-2,10-Camphorsultam may be employed as a chiral probe for the optical resolution by HPLC and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids.[1]

Application

(1S)-(−)-2,10-Camphorsultam may be used in the asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-α-methylcysteine.[2] It may be used as proton source in the synthesis of chiral α,γ-substituted γ-butyrolactones.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Xu et al.
The Journal of organic chemistry, 66(11), 3953-3962 (2001-05-26)
A highly useful method for the synthesis of optically active alpha,gamma-substituted gamma-butyrolactones has been developed. The SmI(2)-induced reductive coupling of chiral 2-alkyl acrylates derived from isosorbide with ketones in the presence of (1S)-(-)-2,10-camphorsultam as a proton source give the chiral
A chiral probe useful for optical resolution and X-ray crystallographic determination of the absolute stereochemistry of carboxylic acids.
Harada N, et al.
Tetrahedron Asymmetry, 4(8), 1755-1758 (1993)
Satendra Singh et al.
The Journal of organic chemistry, 69(13), 4551-4554 (2004-06-19)
(1R)-(+)-2,10- and (1S)-(-)-2,10-camphorsultam were acylated with ethyl 2-phenylthiazoline 4-carboxylate to afford (+)- and (-)-2-phenylthiazolinylcamphorsultam, which were stereoselectively alkylated with MeI in the presence of n-BuLi. Alkylation of these phenylthiazolinylcamphorsultams occurred from the beta-face rather than alpha-face, resulting in the formation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service