Skip to Content
Merck
All Photos(1)

Key Documents

8.02505

Sigma-Aldrich

trans-Cinnamaldehyde

for synthesis

Synonym(s):

trans-Cinnamaldehyde, trans-Cinnamic aldehyde, trans-3-Phenyl-2-propenal

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
€31.40
250 ML
€33.20
1 L
€128.00

€31.40


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 ML
€31.40
250 ML
€33.20
1 L
€128.00

About This Item

Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352114
EC Index Number:
203-213-9
NACRES:
NA.22

€31.40


Estimated to ship on21 May 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2220 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

251 °C/960 hPa

mp

-8 °C

transition temp

flash point 138 °C

density

1.05 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O=C\C=C\c1ccccc1

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.048 - 1.051
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

221.0 °F - Pensky-Martens closed cup

Flash Point(C)

105 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service