Skip to Content
Merck
All Photos(1)

Documents

239968

Sigma-Aldrich

trans-Cinnamaldehyde

≥99%

Synonym(s):

trans-3-Phenyl-2-propenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCHO
CAS Number:
Molecular Weight:
132.16
Beilstein:
1071571
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.6 (vs air)

Assay

≥99%

form

liquid

refractive index

n20/D 1.622 (lit.)

bp

250-252 °C (lit.)

mp

−9-−4 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\c1ccccc1

InChI

1S/C9H8O/c10-8-4-7-9-5-2-1-3-6-9/h1-8H/b7-4+

InChI key

KJPRLNWUNMBNBZ-QPJJXVBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The neuroprotective and anti-inflammatory effects of trans-cinnamaldehyde (TCA) were studied.

Application

trans-Cinnamaldehyde was used to study the useful amino groups at aminosilanized surface.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ji-Hi Pyo et al.
Biological & pharmaceutical bulletin, 36(12), 1928-1935 (2013-12-03)
The anti-inflammatory and neuroprotective effects of trans-cinnamaldehyde (TCA) were investigated on the inflammatory cells and the dopaminergic degeneration in mice. TCA inhibited the up-regulation of inducible nitric oxide synthase (iNOS) and cyclooxygenase-2 (COX-2) in the lipopolysaccharide (LPS)-induced inflammatory BV2 microglial
Wen-Xian Du et al.
Journal of agricultural and food chemistry, 60(32), 7799-7804 (2012-07-27)
The addition of plant essential oils to edible films and coatings has been shown to protect against bacterial pathogens and spoilage while also enhancing sensory properties of foods. This study evaluated the effect of adding 0.5 and 0.75% carvacrol (active
Katherine R Zodrow et al.
Langmuir : the ACS journal of surfaces and colloids, 28(39), 13993-13999 (2012-09-04)
Biofilm-associated infections are one of the leading causes of death in the United States. Although infections may be treated with antibiotics, the overuse of antibiotics has led to the spread of antibiotic resistance. Many natural antimicrobial compounds derived from edible
Amrita A Nagle et al.
PloS one, 7(11), e50125-e50125 (2012-11-28)
Multifunctional trans-cinnamaldehyde (CA) and its analogs display anti-cancer properties, with 2-benzoyloxycinnamaldehyde (BCA) and 5-fluoro-2-hydroxycinnamaldehyde (FHCA) being identified as the ortho-substituted analogs that possess potent anti-tumor activities. In this study, BCA, FHCA and a novel analog 5-fluoro-2-benzoyloxycinnamaldehyde (FBCA), were demonstrated to
Marie E Barabas et al.
PloS one, 7(10), e47988-e47988 (2012-11-08)
Subpopulations of somatosensory neurons are characterized by functional properties and expression of receptor proteins and surface markers. CGRP expression and IB4-binding are commonly used to define peptidergic and non-peptidergic subpopulations. TRPA1 is a polymodal, plasma membrane ion channel that contributes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service